Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Vps33a
Entrez Gene ID
77573
Gene Name
vacuolar protein sorting 33A (yeast)
Gene Aliases
3830421M04Rik, AI503300, AW048546, AW554476, bf
Chromosome Location
Chr.5: 123527304 - 123573043 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_029929.3 NP_084205.3 9 - 10 1209 72
GenBank mRNA AF439858.1 - 9 - 10 1193 72
AK019463.1 - 9 - 10 1193 72
AK041982.1 - 9 - 10 1205 72
AK140553.1 - 9 - 10 1198 72
AK150282.1 - 9 - 10 1201 72
AK150550.1 - 9 - 10 1201 72
AK206082.1 - 1 - 2 81 72
BC046417.1 - 9 - 10 1200 72

View Full Product Details