The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_027456.1 | NP_081732.1 | 9 - 10 | 948 | 94 |
NM_030184.2 | NP_084460.1 | 9 - 10 | 983 | 94 | |
GenBank mRNA | AK017564.1 | - | 9 - 10 | 949 | 94 |
AK019600.1 | - | 9 - 10 | 972 | 94 | |
AK036083.1 | - | 9 - 10 | 954 | 94 | |
AK039800.1 | - | 9 - 10 | 956 | 94 | |
AK045699.1 | - | 9 - 10 | 947 | 94 | |
AK046693.1 | - | 9 - 10 | 966 | 94 | |
AK140598.1 | - | 9 - 10 | 979 | 94 | |
AK161393.1 | - | 9 - 10 | 963 | 94 | |
AK164041.1 | - | 9 - 10 | 982 | 94 | |
BC028964.1 | - | 2 - 3 | 203 | 94 | |
BC043101.1 | - | 9 - 10 | 935 | 94 |