The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001110320.1 | NP_001103790.1 | 2 - 3 | 292 | 58 |
NM_007654.2 | NP_031680.2 | 2 - 3 | 292 | 58 | |
GenBank mRNA | AF543213.1 | - | 2 - 3 | 193 | 58 |
AK134132.1 | - | 2 - 3 | 295 | 58 | |
BC003824.1 | - | 2 - 3 | 250 | 58 | |
J04170.1 | - | 2 - 3 | 229 | 58 | |
S40777.1 | - | 2 - 3 | 225 | 58 |