The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_177406.3 | NP_803125.2 | 6 - 7 | 862 | 76 |
GenBank mRNA | AK144260.1 | - | 6 - 7 | 792 | 76 |
BC014721.1 | - | 6 - 7 | 808 | 76 | |
BC025936.1 | - | 6 - 7 | 802 | 76 | |
BC026582.1 | - | 6 - 7 | 808 | 76 | |
BC031141.1 | - | 6 - 7 | 783 | 76 | |
BC033924.1 | - | 6 - 7 | 792 | 76 |