Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Eps15
Entrez Gene ID
13858
Gene Name
epidermal growth factor receptor pathway substrate 15
Gene Aliases
2410112D09Rik
Chromosome Location
Chr.4: 109280256 - 109387817 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001159964.1 NP_001153436.1 3 - 4 663 102
NM_007943.3 NP_031969.1 15 - 16 1612 102
GenBank mRNA AK028361.1 - - 811 102
AK083176.1 - 3 - 4 647 102
AY190610.1 - 15 - 16 1585 102
BC048783.1 - 15 - 16 1586 102
L21768.1 - 15 - 16 1585 102

View Full Product Details