Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ercc2
Entrez Gene ID
13871
Gene Name
excision repair cross-complementing rodent repair deficiency, complementation group 2
Gene Aliases
AA407812, AU020867, AW240756, CXPD, Ercc-2, XPD
Chromosome Location
Chr.7: 19382039 - 19395692 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_007949.4 NP_031975.2 6 - 7 490 54
GenBank mRNA AK004652.1 - 2 - 3 142 54
AK082761.1 - 6 - 7 491 54
AK141692.1 - 6 - 7 509 54
AK145969.1 - 6 - 7 499 54
BC034517.1 - 5 - 6 426 54
U97572.1 - 5 - 6 474 54

View Full Product Details