Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ercc2
Entrez Gene ID
13871
Gene Name
excision repair cross-complementing rodent repair deficiency, complementation group 2
Gene Aliases
AA407812, AU020867, AW240756, CXPD, Ercc-2, XPD
Chromosome Location
Chr.7: 19382039 - 19395692 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_007949.4 NP_031975.2 18 - 19 1773 84
GenBank mRNA AK004652.1 - 14 - 15 1425 84
AK082323.1 - 11 - 12 1027 85
AK082761.1 - 18 - 19 1774 84
AK145969.1 - 18 - 19 1782 84
AK209753.1 - 3 - 4 237 84
BC034517.1 - 17 - 18 1709 84
U97572.1 - 17 - 18 1757 84

View Full Product Details