The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_007989.4 | NP_032015.1 | 1 - 2 | 347 | 79 |
GenBank mRNA | AF069303.1 | - | 1 - 2 | 273 | 79 |
AF079514.1 | - | 1 - 2 | 270 | 79 | |
AF110506.1 | - | 1 - 2 | 270 | 79 | |
AF177770.1 | - | 1 - 2 | 270 | 79 | |
BC119156.1 | - | 1 - 2 | 347 | 79 | |
BC137781.1 | - | 1 - 2 | 347 | 79 |