The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_021316.4 | NP_067291.1 | 1 - 2 | 109 | 84 |
GenBank mRNA | AF243130.1 | - | 1 - 2 | 46 | 84 |
AK079045.1 | - | 1 - 2 | 110 | 84 | |
BC023032.1 | - | 1 - 2 | 47 | 84 |