Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Angel2
Entrez Gene ID
52477
Gene Name
angel homolog 2 (Drosophila)
Gene Aliases
2610307I21Rik, 5730410O10Rik, AI845692, D1Ertd396e, D1Ertd654e
Chromosome Location
Chr.1: 190925108 - 190946962 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001199020.1 NP_001185949.1 5 - 6 766 75
NM_021421.4 NP_067396.3 4 - 5 901 75
NR_037578.1 4 - 5 860 75
NR_037579.1 3 - 4 575 75
GenBank mRNA AB041602.1 - 3 - 4 561 75
AK077039.1 - 4 - 5 847 75
AK088834.1 - 3 - 4 371 75
AK135709.1 - 5 - 6 763 75
AK158731.1 - 4 - 5 917 75
AK166619.1 - 5 - 6 765 75
AK169653.1 - 3 - 4 485 75
AK206765.1 - 2 - 3 149 75
BC024907.1 - 4 - 5 861 75

View Full Product Details