The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_021719.4 | NP_068365.1 | 1 - 2 | 454 | 82 |
GenBank mRNA | AF124427.1 | - | 1 - 2 | 216 | 82 |
AK008227.1 | - | 1 - 2 | 252 | 82 | |
AK008683.1 | - | 1 - 2 | 455 | 82 | |
BC023428.1 | - | 1 - 2 | 443 | 82 |