The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_024223.2 | NP_077185.1 | 6 - 7 | 896 | 79 |
GenBank mRNA | AF469648.1 | - | 6 - 7 | 602 | 79 |
AK002484.1 | - | 6 - 7 | 581 | 79 | |
BC002093.1 | - | 6 - 7 | 599 | 79 | |
BC002096.1 | - | 6 - 7 | 582 | 79 | |
BC007149.1 | - | 6 - 7 | 582 | 79 | |
CT010309.1 | - | 6 - 7 | 560 | 79 |