The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_025840.3 | NP_080116.2 | 6 - 7 | 709 | 75 |
GenBank mRNA | AB244988.1 | - | 6 - 7 | 625 | 75 |
AK003229.1 | - | 6 - 7 | 704 | 75 | |
AK011253.1 | - | 6 - 7 | 665 | 75 | |
AK132595.1 | - | 6 - 7 | 651 | 75 | |
AK160544.1 | - | 6 - 7 | 682 | 75 | |
AK162647.1 | - | 6 - 7 | 651 | 75 | |
BC013060.1 | - | 6 - 7 | 645 | 75 |