Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Klhl7
Entrez Gene ID
52323
Gene Name
kelch-like 7
Gene Aliases
2700038B03Rik, D5Ertd363e, SBBI26
Chromosome Location
Chr.5: 24100574 - 24163365 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_026448.3 NP_080724.2 9 - 10 1545 88
GenBank mRNA AK012326.1 - 9 - 10 1526 88
AK083358.1 - 9 - 10 1531 88
AK089072.1 - 10 - 11 1652 88

View Full Product Details