Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Fktn
Entrez Gene ID
246179
Gene Name
fukutin
Gene Aliases
D830030O17Rik, Fcmd
Chromosome Location
Chr.4: 53714107 - 53765785 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_139309.4 NP_647470.1 5 - 6 893 78
GenBank mRNA AB077383.1 - 6 - 7 1077 78
AJ511807.1 - 6 - 7 913 78
AK085931.1 - 5 - 6 894 78
AK142744.1 - - 1053 78
BC017538.1 - 6 - 7 909 78

View Full Product Details