Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Dus3l
Entrez Gene ID
224907
Gene Name
dihydrouridine synthase 3-like (S. cerevisiae)
Gene Aliases
AI662135, AW557805
Chromosome Location
Chr.17: 56764717 - 56770093 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_144858.2 NP_659107.2 12 - 13 1940 58
GenBank mRNA AA123957.1 - 2 - 3 234 58
AK157702.1 - 12 - 13 1881 58
AK164614.1 - 9 - 10 1821 58
AK191282.1 - 2 - 3 161 58
AK217151.1 - 2 - 3 161 58
AY040840.1 - 12 - 13 1913 58
BC023942.1 - 6 - 7 1490 58
BC052481.1 - 9 - 10 2385 58
BC053941.1 - 9 - 10 2385 58
BC054069.1 - 9 - 10 2385 58
BM224665.2 - 1 - 2 58

View Full Product Details