Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Tmc6
Entrez Gene ID
217353
Gene Name
transmembrane channel-like gene family 6
Gene Aliases
D11Ertd204e, EVER1
Chromosome Location
Chr.11: 117765985 - 117782193 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_145439.2 NP_663414.3 17 - 18 2418 63
GenBank mRNA AK077671.1 - 15 - 16 2163 63
AK090179.1 - 17 - 18 2373 63
AY236497.1 - 17 - 18 2317 63
AY263158.1 - 17 - 18 2343 63
BC004840.1 - 17 - 18 2343 63
BC013502.1 - 8 - 9 1145 63
BC058195.1 - 17 - 18 2343 63
BP767442.1 - 3 - 4 63

View Full Product Details