Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Lhx1
Entrez Gene ID
16869
Gene Name
LIM homeobox protein 1
Gene Aliases
Lim1
Chromosome Location
Chr.11: 84519379 - 84525534 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_008498.2 NP_032524.1 4 - 5 2197 55
GenBank mRNA AK032067.1 - - 388 55
AK141086.1 - - 2199 55
BC092374.1 - 4 - 5 2330 55
S68107.1 - 7 - 8 1729 55
Z27410.1 - 4 - 5 2084 55

View Full Product Details