Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Trub2
Entrez Gene ID
227682
Gene Name
TruB pseudouridine (psi) synthase homolog 2 (E. coli)
Gene Aliases
G430055L02Rik
Chromosome Location
Chr.2: 29761223 - 29787810 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001290495.1 NP_001277424.1 4 - 5 459 77
NM_001290496.1 NP_001277425.1 5 - 6 521 77
NM_145520.5 NP_663495.3 5 - 6 521 77
NR_110962.1 4 - 5 459 77
GenBank mRNA AK043949.1 - 5 - 6 516 77
AK050587.1 - 4 - 5 449 77
AK081579.1 - 4 - 5 449 77
AK087102.1 - 5 - 6 503 77
AK090004.1 - - 1353 77
AK167294.1 - 5 - 6 497 77
BC015285.1 - 5 - 6 500 77
BC024075.1 - 5 - 6 490 77
BU053791.1 - 5 - 6 473 77
BU707805.1 - 4 - 5 421 77

View Full Product Details