The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_145974.3 | NP_666086.2 | 1 - 2 | 395 | 68 |
GenBank mRNA | AK049249.1 | - | 1 - 2 | 165 | 68 |
AK077848.1 | - | 1 - 2 | 158 | 68 | |
AK142500.1 | - | 1 - 2 | 158 | 68 | |
AK170270.1 | - | 1 - 2 | 193 | 68 | |
AK170521.1 | - | 1 - 2 | 190 | 68 | |
BC006740.1 | - | 1 - 2 | 108 | 68 | |
BC055831.1 | - | 1 - 2 | 395 | 68 |