The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_145997.2 | NP_666109.2 | 11 - 12 | 2005 | 130 |
GenBank mRNA | AK144877.1 | - | 11 - 12 | 1976 | 130 |
AK159079.1 | - | 11 - 12 | 1982 | 130 | |
AK166055.1 | - | 11 - 12 | 1986 | 130 | |
BC010717.1 | - | 11 - 12 | 1646 | 130 | |
BC011506.3 | - | 11 - 12 | 1939 | 130 | |
BC080691.1 | - | 11 - 12 | 1691 | 130 |