Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ermp1
Entrez Gene ID
226090
Gene Name
endoplasmic reticulum metallopeptidase 1
Gene Aliases
D19Ertd410e, D19Wsu12e, Fxna, mKIAA1815
Chromosome Location
Chr.19: 29609883 - 29648484 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001081213.1 NP_001074682.1 10 - 11 1963 93
GenBank mRNA AK050284.1 - 10 - 11 1959 93
AK081674.1 - 9 - 10 1480 93
AK129447.1 - 10 - 11 1891 93
AK137206.1 - 10 - 11 1919 93
BC031519.1 - 2 - 3 239 93
BC086627.1 - 10 - 11 1904 93
BC151181.1 - 10 - 11 1922 93
BC151182.1 - 10 - 11 1922 93
BC157914.1 - 10 - 11 1922 93

View Full Product Details