Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Slc29a4
Entrez Gene ID
243328
Gene Name
solute carrier family 29 (nucleoside transporters), member 4
Gene Aliases
ENT4, mPMAT
Chromosome Location
Chr.5: 142692476 - 142722490 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_146257.2 NP_666369.1 9 - 10 1403 80
GenBank mRNA BC025599.1 - 9 - 10 1347 80

View Full Product Details