The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_019440.3 | NP_062313.3 | 2 - 2 | 1249 | 68 |
GenBank mRNA | AJ007972.1 | - | 1 - 1 | 1050 | 68 |
AK142291.1 | - | 2 - 2 | 1251 | 68 | |
AK152870.1 | - | 2 - 2 | 1190 | 68 | |
AK153309.1 | - | 2 - 2 | 1190 | 68 | |
AK165347.1 | - | 2 - 2 | 1187 | 68 |