The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_172116.4 | NP_742114.3 | 3 - 4 | 312 | 58 |
GenBank mRNA | AK081187.1 | - | 3 - 4 | 272 | 58 |
AK084273.1 | - | 3 - 4 | 268 | 58 | |
AK088159.1 | - | 3 - 4 | 288 | 58 | |
AK140975.1 | - | 3 - 4 | 263 | 58 | |
AK161205.1 | - | 3 - 4 | 278 | 58 | |
BC023835.1 | - | 3 - 4 | 282 | 58 |