The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_012056.2 | NP_036186.2 | 9 - 10 | 1686 | 67 |
GenBank mRNA | AF090334.1 | - | 9 - 10 | 1612 | 67 |
AF279263.1 | - | 9 - 10 | 1664 | 67 | |
AK007499.1 | - | 7 - 8 | 1076 | 67 | |
AK212595.1 | - | 1 - 2 | 97 | 67 | |
AK218657.1 | - | 2 - 3 | 295 | 67 | |
BC026133.1 | - | 9 - 10 | 1626 | 67 | |
BC043129.1 | - | 9 - 10 | 1686 | 67 |