The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_027301.3 | NP_081577.1 | 1 - 2 | 410 | 61 |
GenBank mRNA | AK037184.1 | - | 1 - 2 | 428 | 61 |
AY044433.1 | - | 1 - 2 | 383 | 61 | |
BC064820.1 | - | 1 - 2 | 410 | 61 |