The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_028041.2 | NP_082317.1 | 1 - 2 | 174 | 92 |
GenBank mRNA | AF319547.1 | - | 1 - 2 | 534 | 92 |
AK028274.1 | - | 1 - 2 | 188 | 92 | |
AK031534.1 | - | 1 - 2 | 184 | 92 | |
AK031679.1 | - | 1 - 2 | 192 | 92 | |
AK147715.1 | - | 1 - 2 | 189 | 92 | |
AK152989.1 | - | 1 - 2 | 188 | 92 | |
AK155138.1 | - | 1 - 2 | 186 | 92 | |
AK167603.1 | - | 1 - 2 | 188 | 92 | |
AK168455.1 | - | 1 - 2 | 188 | 92 | |
AK169255.1 | - | 1 - 2 | 186 | 92 | |
AK170967.1 | - | 1 - 2 | 186 | 92 | |
AK216814.1 | - | - | 279 | 92 | |
BC043699.1 | - | 1 - 2 | 174 | 92 | |
BC062920.1 | - | 1 - 2 | 177 | 92 |