The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_172490.3 | NP_766078.1 | 1 - 2 | 129 | 62 |
GenBank mRNA | AK033654.1 | - | 1 - 2 | 130 | 62 |
AK049338.1 | - | - | 136 | 62 | |
AK153792.1 | - | - | 136 | 62 | |
BC062133.1 | - | 1 - 2 | 119 | 62 |