The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_029885.1 | NP_084161.1 | 4 - 5 | 643 | 70 |
NM_172512.2 | NP_766100.1 | 3 - 4 | 546 | 70 | |
GenBank mRNA | AK030849.1 | - | 3 - 4 | 315 | 70 |
AK036692.1 | - | 4 - 5 | 411 | 70 | |
AK086817.1 | - | 3 - 4 | 490 | 70 | |
AK087979.1 | - | 4 - 5 | 460 | 70 | |
AK141382.1 | - | 4 - 5 | 617 | 70 | |
AK163939.1 | - | 4 - 5 | 405 | 70 | |
BC150160.1 | - | 4 - 5 | 394 | 70 | |
BC150482.1 | - | 4 - 5 | 394 | 70 |