Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Zbbx
Entrez Gene ID
213234
Gene Name
zinc finger, B-box domain containing
Gene Aliases
4931432L23
Chromosome Location
Chr.3: 75037895 - 75165050 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_172515.2 NP_766103.2 16 - 17 2236 99
GenBank mRNA AK029899.1 - 16 - 17 2229 99
BC050789.1 - 16 - 17 2236 99

View Full Product Details