Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Nutm1
Entrez Gene ID
213765
Gene Name
NUT midline carcinoma, family member 1
Gene Aliases
4932438M10, Nut
Chromosome Location
Chr.2: 112247948 - 112259291 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_172521.1 NP_766109.1 1 - 2 125 79
GenBank mRNA AK077049.1 - 1 - 2 125 79
BC125332.1 - 1 - 2 105 79
BC125334.1 - 1 - 2 105 79

View Full Product Details