The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001164612.1 | NP_001158084.1 | 14 - 15 | 1851 | 64 |
NM_001164613.1 | NP_001158085.1 | 15 - 16 | 1904 | 64 | |
NM_172613.4 | NP_766201.3 | 14 - 15 | 1794 | 64 | |
GenBank mRNA | AK028776.1 | - | 15 - 16 | 1906 | 64 |
AK029303.1 | - | 13 - 14 | 1595 | 64 |