The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001080754.1 | NP_001074223.1 | 9 - 10 | 2247 | 68 |
NM_172669.3 | NP_766257.3 | 8 - 9 | 2520 | 68 | |
GenBank mRNA | AK083596.1 | - | 8 - 9 | 2477 | 68 |
AK084557.1 | - | 8 - 9 | 2502 | 68 | |
AK129433.1 | - | 4 - 5 | 1784 | 68 | |
AK155513.1 | - | 8 - 9 | 2483 | 68 | |
AK169280.1 | - | 9 - 10 | 2200 | 68 | |
BC032293.1 | - | 9 - 10 | 2170 | 68 | |
BC054082.1 | - | 8 - 9 | 2452 | 68 |