The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_172689.3 | NP_766277.3 | 15 - 16 | 2317 | 72 |
GenBank mRNA | AK049305.1 | - | 6 - 7 | 908 | 72 |
AK052595.1 | - | 11 - 12 | 1571 | 72 | |
AK128929.1 | - | 15 - 16 | 2252 | 72 | |
AY553221.1 | - | 15 - 16 | 2193 | 72 | |
BC027369.1 | - | 15 - 16 | 2296 | 72 | |
BC106095.1 | - | 15 - 16 | 2237 | 72 | |
BC127163.1 | - | 15 - 16 | 2183 | 72 | |
BC152540.1 | - | 15 - 16 | 2185 | 72 |