Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Dok7
Entrez Gene ID
231134
Gene Name
docking protein 7
Gene Aliases
A930013K19Rik, AW049091, Dok-7, EF-12, Oit5
Chromosome Location
Chr.5: 35056566 - 35087839 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_172708.3 NP_766296.1 1 - 2 56 85
GenBank mRNA AK044445.1 - 1 - 2 57 85

View Full Product Details