The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001110827.2 | NP_001104297.1 | 13 - 14 | 1801 | 87 |
NM_001110828.2 | NP_001104298.1 | 12 - 13 | 1355 | 87 | |
NM_001110829.2 | NP_001104299.1 | 12 - 13 | 1346 | 87 | |
NM_001110830.2 | NP_001104300.1 | 12 - 13 | 1343 | 87 | |
NM_001286418.1 | NP_001273347.1 | 9 - 10 | 1061 | 87 | |
NM_001286419.1 | NP_001273348.1 | 11 - 12 | 1250 | 87 | |
NM_053104.6 | NP_444334.3 | 13 - 14 | 1813 | 87 | |
NM_175387.3 | NP_780596.1 | 12 - 13 | 1761 | 87 | |
GenBank mRNA | AF229055.1 | - | 12 - 13 | 1286 | 87 |
AK077594.1 | - | 13 - 14 | 1689 | 87 | |
AK139555.1 | - | 14 - 15 | 1785 | 87 | |
BC002124.1 | - | 12 - 13 | 1181 | 87 | |
BC027263.1 | - | 12 - 13 | 1313 | 87 |