The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_173785.6 | NP_776146.1 | 1 - 2 | 393 | 78 |
GenBank mRNA | AK038625.1 | - | 1 - 2 | 388 | 78 |
AK133203.1 | - | 1 - 2 | 391 | 78 | |
AK172525.1 | - | 1 - 2 | 354 | 78 | |
BC094909.1 | - | 1 - 2 | 393 | 78 |