The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_032000.2 | NP_114389.2 | 6 - 6 | 8650 | 74 |
GenBank mRNA | AK046585.1 | - | 1 - 1 | 3155 | 74 |
AK078617.1 | - | 1 - 1 | 1120 | 74 |