Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cntln
Entrez Gene ID
338349
Gene Name
centlein, centrosomal protein
Gene Aliases
AI448092, B430108F07Rik, D530005L17Rik
Chromosome Location
Chr.4: 84883512 - 85131923 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_175275.4 NP_780484.2 1 - 2 517 71
NM_177385.4 NP_796359.1 1 - 2 517 71
GenBank mRNA AK031604.1 - 1 - 2 423 71
AK052558.1 - 1 - 2 437 71
BC125622.1 - 1 - 2 392 71
BC125624.1 - 1 - 2 392 71
BM942195.1 - 1 - 2 517 71

View Full Product Details