Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ofd1
Entrez Gene ID
237222
Gene Name
oral-facial-digital syndrome 1 gene homolog (human)
Gene Aliases
Cxorf5, DXGgc7e, ORF2
Chromosome Location
Chr.X: 166390033 - 166440838 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_177429.3 NP_803178.2 14 - 15 1956 61
GenBank mRNA AJ438159.1 - 14 - 15 1807 61
BC119070.1 - 14 - 15 1639 61
BC120487.1 - 14 - 15 1639 61

View Full Product Details