Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Wdr36
Entrez Gene ID
225348
Gene Name
WD repeat domain 36
Gene Aliases
5730444A13Rik, Ta-wdrp
Chromosome Location
Chr.18: 32837225 - 32866420 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001110015.1 NP_001103485.1 11 - 12 1353 66
NM_001110016.1 NP_001103486.1 11 - 12 1353 66
NM_144863.4 NP_659112.2 11 - 12 1353 66
GenBank mRNA AK041763.1 - 11 - 12 1273 66
AK045396.1 - 11 - 12 1326 66
AK077545.1 - 11 - 12 1282 66
AK155965.1 - 11 - 12 1251 66
AK156822.1 - - 1354 66
AK171683.1 - 11 - 12 1291 66
AK172054.1 - 11 - 12 1278 66
AK172377.1 - 11 - 12 1283 66
BC022132.1 - 11 - 12 1325 66
BC025809.1 - 11 - 12 1267 66
BC094453.1 - 11 - 12 1304 66

View Full Product Details