The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001293560.1 | NP_001280489.1 | 11 - 12 | 1269 | 96 |
NM_173763.4 | NP_776124.1 | 11 - 12 | 1273 | 96 | |
GenBank mRNA | AF363737.1 | - | 11 - 12 | 1243 | 96 |
AK049569.1 | - | 11 - 12 | 1242 | 96 | |
AK145623.1 | - | 11 - 12 | 1267 | 96 | |
BC037135.1 | - | 7 - 8 | 850 | 96 | |
BC085287.1 | - | 12 - 13 | 1366 | 96 | |
BC131942.1 | - | 10 - 11 | 1058 | 96 | |
BC132615.1 | - | 10 - 11 | 1058 | 96 |