Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Fam53b
Entrez Gene ID
77938
Gene Name
family with sequence similarity 53, member B
Gene Aliases
A930008G19Rik, mKIAA0140
Chromosome Location
Chr.7: 132712078 - 132813196 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_175268.4 NP_780477.2 3 - 4 372 98
NM_212473.1 NP_997638.1 3 - 4 435 98
GenBank mRNA AK020830.1 - 3 - 4 373 98
AK031576.1 - 2 - 3 216 98
AK051299.1 - 3 - 4 436 98
AK147350.1 - 3 - 4 768 98
AK147725.1 - 3 - 4 564 98
AK156629.1 - 3 - 4 335 98
AK187644.1 - 2 - 3 97
BC035523.1 - 3 - 4 367 98

View Full Product Details