Product Details
- Assay Reference Genome
Location
Chr.17:55956681 on build GRCm38
- Cytoband
- 17D
- Species
- Mus musculus
- Variation Type
- Copy Number
Additional Information
Target Gene Details
- Entrez Gene ID
- 50498
- Gene Symbol
- Ebi3
- Gene Name
- Epstein-Barr virus induced gene 3
- Gene Aliases
- EBI-3, IL-27
- Location
- Chr.17:55952623-55957022 on build GRCm38
- Assay Gene Location
- Within Exon 5
- Entrez Gene ID
- 100504333
- Gene Symbol
- Gm16712
- Gene Name
- predicted gene, 16712
- Gene Aliases
- Location
- Chr.17:55954771-55959401 on build GRCm38
- Assay Gene Location
- Within Intron 2
View Full Product Details