Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Cyp4f39
Entrez Gene ID
320997
Gene Name
cytochrome P450, family 4, subfamily f, polypeptide 39
Gene Aliases
4732474A20Rik
Chromosome Location
Chr.17: 32452695 - 32493320 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_177307.3 NP_796281.1 12 - 13 1514 65
GenBank mRNA AK028950.1 - 12 - 13 1515 65
AK037192.1 - 12 - 13 1515 65
BC145755.1 - 12 - 13 1511 65

View Full Product Details