Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ccdc60
Entrez Gene ID
269693
Gene Name
coiled-coil domain containing 60
Gene Aliases
AU018954, C130098D09
Chromosome Location
Chr.5: 116125581 - 116288998 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_177759.3 NP_808427.1 12 - 13 1791 78
GenBank mRNA AK082047.1 - 12 - 13 1792 78
BC060151.1 - - 306 78
BC150892.1 - 12 - 13 1581 78
BC172040.1 - 12 - 13 1579 78

View Full Product Details