Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Srsf12
Entrez Gene ID
272009
Gene Name
serine/arginine-rich splicing factor 12
Gene Aliases
B830039L16, Sfrs13b, Srrp, Srsf13b
Chromosome Location
Chr.4: 33208987 - 33233344 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_177774.4 NP_808442.2 1 - 2 378 107
GenBank mRNA AK046869.1 - 1 - 2 379 107

View Full Product Details