The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_178775.4 | NP_848890.3 | 10 - 11 | 1336 | 97 |
GenBank mRNA | AK044809.1 | - | 7 - 8 | 1055 | 97 |
AK049003.1 | - | 3 - 4 | 284 | 97 | |
AK149858.1 | - | 10 - 11 | 1301 | 97 |