Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Arrdc3
Entrez Gene ID
105171
Gene Name
arrestin domain containing 3
Gene Aliases
AI450344, mKIAA1376
Chromosome Location
Chr.13: 80883338 - 80896042 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001042591.1 NP_001036056.1 1 - 2 507 125
GenBank mRNA AK084951.1 - 1 - 2 518 125
AK088288.1 - 1 - 2 522 125
AK122499.1 - 1 - 2 494 125
BC044762.1 - 1 - 2 505 125
BC054844.1 - 1 - 2 507 125
BC098474.1 - 1 - 2 501 125
DQ861994.1 - 1 - 2 252 125

View Full Product Details